Hydrochloric acid, 6.0N Standardized Solution
Hydrochloric acid, 6.0N Standardized Solution
Hydrochloric acid, 6.0N Standardized Solution
Thermo Scientific Chemicals

Hydrochloric acid, 6.0N Standardized Solution

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
20 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044921.K7
also known as 044921-K7
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration6.0N Hydrochloric acid
MatrixWater
6.0N HClsolution acts as a standard solution in volumetric titrations. In addition, it has industrial applications such as chemical transformations in hydrochlorination of rubber and production of vinyl and alkyl halide. In the semiconductor industry, it is used to etch semiconductor crystal and to purify silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only