Holmium(III) fluoride, anhydrous, 99.9% (REO)
Holmium(III) fluoride, anhydrous, 99.9% (REO)
Holmium(III) fluoride, anhydrous, 99.9% (REO)
Thermo Scientific Chemicals

Holmium(III) fluoride, anhydrous, 99.9% (REO)

CAS: 13760-78-6 | F3Ho | 221.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 045033.09
also known as 045033-09
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS13760-78-6
IUPAC Nameholmium(3+) trifluoride
Molecular FormulaF3Ho
InChI KeyFDIFPFNHNADKFC-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) fluoride, anhydrous is used to produce the strongest artificially- generated magnetic fields, nuclear control rods, yellow-colored glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only