Sulfuric acid, 0.05N Standardized Solution
Sulfuric acid, 0.05N Standardized Solution
Sulfuric acid, 0.05N Standardized Solution
Thermo Scientific Chemicals

Sulfuric acid, 0.05N Standardized Solution

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 045048.K7
亦稱為 045048-K7
價格 (TWD)
-
Quantity:
4 L
Request bulk or custom format
化學識別
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
FormLiquid
Concentration0.0500N ± 0.21%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only