Platinum foil, 0.203mm (0.008in) thick, 99.9% (metals basis)
Platinum foil, 0.203mm (0.008in) thick, 99.9% (metals basis)
Platinum foil, 0.203mm (0.008in) thick, 99.9% (metals basis)
Platinum foil, 0.203mm (0.008in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum foil, 0.203mm (0.008in) thick, 99.9% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
045091.FF
also known as 045091-FF
25 x 25 mm
Catalog number 045091.FF
also known as 045091-FF
Price (USD)
833.65
Online Exclusive
926.00
Save 92.35 (10%)
Each
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
833.65
Online Exclusive
926.00
Save 92.35 (10%)
Each
Add to cart
Specifications
Assay Percent Notes(metals basis)
CAS7440-06-4
ChEBICHEBI:33400
Chemical Name or MaterialPlatinum foil
EINECS Number231-116-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only