Selenium ingot/button,≈38mm (1.5 in.) dia.
Selenium ingot/button,≈38mm (1.5 in.) dia.
Selenium ingot/button,≈38mm (1.5 in.) dia.
Selenium ingot/button,≈38mm (1.5 in.) dia.
Thermo Scientific Chemicals

Selenium ingot/button,≈38mm (1.5 in.) dia.

CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Catalog NumberQuantity
045115.KF
also known as 045115-KF
1 Pc.
Catalog number 045115.KF
also known as 045115-KF
Price (TWD)
-
Request A Quote
Quantity:
1 Pc.
Request bulk or custom format
Specifications
CAS7782-49-2
ChEBICHEBI:27568
Chemical Name or MaterialSelenium ingot/button
DOT InformationHazard Class: 6.1; Packaging Group: III
EINECS Number231-957-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only