Iridium slug, 6.0mm (0.24in) dia x 12.0mm (0.48in) length, 99.8% min. (metals basis)
Iridium slug, 6.0mm (0.24in) dia x 12.0mm (0.48in) length, 99.8% min. (metals basis)
Iridium slug, 6.0mm (0.24in) dia x 12.0mm (0.48in) length, 99.8% min. (metals basis)
Iridium slug, 6.0mm (0.24in) dia x 12.0mm (0.48in) length, 99.8% min. (metals basis)
Thermo Scientific Chemicals

Iridium slug, 6.0mm (0.24in) dia x 12.0mm (0.48in) length, 99.8% min. (metals basis)

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Catalog NumberQuantity
045121.KU5 Ea.
Catalog number 045121.KU
Price (USD)
-
Quantity:
5 Ea.
Request bulk or custom format
Specifications
Chemical Name or MaterialIridium slug
Name Note6.0 mm (0.24 in.) dia. x 12.0 mm (0.48 in.) length
CAS7439-88-5
Recommended StorageAmbient temperatures
EINECS Number231-095-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only