Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals
Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals
Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals

 
Have Questions?
產品號碼Quantity
045324.22
亦稱為 045324-22
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 045324.22
亦稱為 045324-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
規格
CAS540-84-1
ChEBICHEBI:62805
Chemical Name or MaterialLead in Isooctane standard solution
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Health Hazard 1H225-H304-H315-H319-H336
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only