Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
045524.KT
also known as 045524-KT
1 Ea.
Catalog number 045524.KT
also known as 045524-KT
Price (USD)/ Each
296.65
Special offer
Online exclusive
349.00
Save 52.35 (15%)
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
296.65
Special offer
Online exclusive
349.00
Save 52.35 (15%)
Add to cart
Specifications
Assay Percent Notes(metals basis)
CAS7440-15-5
ChEBICHEBI:49882
Chemical Name or MaterialRhenium slug
EINECS Number231-124-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only