Thermo Scientific Chemicals

Gibberellic acid, 98%, Thermo Scientific Chemicals

Catalog number: 045569.04
2 g, Each
Thermo Scientific Chemicals

Gibberellic acid, 98%, Thermo Scientific Chemicals

Catalog number: 045569.04
2 g, Each
Quantity
Catalog number: 045569.04
also known as 045569-04
Price (USD)
Price: 91.50
Online price: 77.65
Your price:
Quantity
-

Chemical Identifiers

CAS
77-06-5
IUPAC Name
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular Formula
C19H22O6
InChI Key
IXORZMNAPKEEDV-QTWFBFKQSA-N
SMILES
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Form
Crystalline powder
Proton NMR
Corresponds to assigned structure
Appearance (Color)
White to very pale cream
Assay (Aqueous acid-base Titration)
≥97.5%
Optical Rotation
76.0 to +86.0°

Description

Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text