Gibberellic acid, 98%
Gibberellic acid, 98%
Gibberellic acid, 98%
Thermo Scientific Chemicals

Gibberellic acid, 98%

Plant growth hormone | CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
10 g
Catalog number 045569.ME
also known as 045569-ME
Price (EUR)
43,70
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
43,70
Each
Chemical Identifiers
CAS77-06-5
IUPAC Name(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular FormulaC19H22O6
InChI KeyIXORZMNAPKEEDV-QTWFBFKQSA-N
SMILESCC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to very pale cream
FormCrystalline powder
Assay from Suppliers CofA≥97.5%
Optical Rotation74.0 to +92.0°
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only