Thermo Scientific Chemicals

Bismuth(III) oxide, NanoArc™, 99.5+%, Thermo Scientific Chemicals

Catalog number: 045582.36
500 g, Each
Thermo Scientific Chemicals

Bismuth(III) oxide, NanoArc™, 99.5+%, Thermo Scientific Chemicals

Catalog number: 045582.36
500 g, Each
Quantity
Catalog number: 045582.36
also known as 045582-36
Price (USD)
Price: 547.00
Your price:
Quantity
-

Chemical Identifiers

CAS
1304-76-3
IUPAC Name
dibismuth(3+) trioxidandiide
Molecular Formula
Bi2O3
InChI Key
TYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Bi+3].[Bi+3]
Assay (unspecified)
99.5% min
Surface Area
12-24 m²/g

Description

Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • X-ray opaque particle
  • NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text