Thermo Scientific™

Misch Metal, 99.0% min. rare earth content, Thermo Scientific™

2 kg, Each
Thermo Scientific™

Misch Metal, 99.0% min. rare earth content, Thermo Scientific™

2 kg, Each
Quantity
Catalog number: 045589.A3
also known as 045589-A3
Price (USD)
1,376.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
62379-61-7
Appearance (Color)
Grey to black
Form
Chunks
Assay from Supplier's CofA
≥99.0% (RE)

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures