Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 1,200, 99%

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 045594.22
also known as 045594-22
Price (USD)
147.65
Online Exclusive
164.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
147.65
Online Exclusive
164.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Assay from Supplier's CofA≥98.5%
FormLiquid
Viscosity3500-7500 cPs
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only