Lead powder, -100 mesh, 99.95% (metals basis)
Lead powder, -100 mesh, 99.95% (metals basis)
Lead powder, -100 mesh, 99.95% (metals basis)
Lead powder, -100 mesh, 99.95% (metals basis)
Thermo Scientific Chemicals

Lead powder, -100 mesh, 99.95% (metals basis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 045602.A1
also known as 045602-A1
Price (USD)
87.65
Special offer
Online exclusive
103.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
87.65
Special offer
Online exclusive
103.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only