Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis) - FAQs
View additional product information for Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis) - FAQs (045603.14, 045603.36, 045603.22)
1 product FAQs found
What is the difference between Titanium (IV) oxide, rutile and Titanium (IV) oxide, anatase?
Rutile and anatase are polymorphs of TiO2, which are both naturally occurring. Rutile is a source for titanium smelting and is used in the semiconductor industry to produce dye-based solar cells (Grätzel cells). Anatase shows a high photocatalytic activity, which limits the usability as a pigment in polymers, but also opens certain applications as a catalyst. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.