Specifications
Avg. Mol. Wt. or Mol. Wt. RangeNi:Fe:Mo; 81:17:2 wt%
Chemical Name or MaterialNickel Iron Molybdenum flake
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Density0.29g/mL
Health Hazard 1H228-H317-H350-H372
View more