Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Thermo Scientific Chemicals

Silicon(IV) nitride, amorphous, nanopowder, 98.5+%

CAS: 12033-89-5 | N4Si3 | 140.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 045611.06
also known as 045611-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS12033-89-5
IUPAC NameSilicon nitride
Molecular FormulaN4Si3
InChI KeyHQVNEWCFYHHQES-UHFFFAOYSA-N
SMILESN12[Si]34N5[Si]11N3[Si]25N41
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only