Thermo Scientific Chemicals

Hexanes, mixed isomers, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 045652.K7
4 L, Each
Thermo Scientific Chemicals

Hexanes, mixed isomers, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 045652.K7
4 L, Each
Quantity
Catalog number: 045652.K7
also known as 045652-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3, 96-14-0, 107-83-5, 96-37-7, 71-43-2
Optical Absorbance at
300nm: ≤0.005AU
Residue on Evaporation
≤5 mg/ml
Assay (GC)
≥60.0% as n-Hexane
Assay (GC)
≥99%, as C6 Hydrocarbons
Optical Absorbance at
195nm: ≤1.00AU

Description

Hexanes, mixed isomers are used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text