Thermo Scientific Chemicals
Nitinol tension coil spring, Alloy M, Thermo Scientific Chemicals
Catalog number: 045655.KM
25 Pc., Each
Thermo Scientific Chemicals
Nitinol tension coil spring, Alloy M, Thermo Scientific Chemicals
Catalog number: 045655.KM
25 Pc., Each
Quantity
Catalog number: 045655.KM
also known as 045655-KM
Price (USD)
Price: 485.00
Online price: 412.65
Your price:
Quantity
-
Chemical Identifiers
CAS
7440-02-0, 7440-32-6
Transition Temperature
approx. 62.5 ?C
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov