Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex

CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 045782.A1
also known as 045782-A1
Price (USD)
531.65
Special offer
Online exclusive
625.00
Save 93.35 (15%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
531.65
Special offer
Online exclusive
625.00
Save 93.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS25655-41-8
Molecular Formula(C6H9NO)nI2
SMILESII.*-CC(-*)N1CCCC1=O
MDL NumberMFCD00084483
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Assay (unspecified)%Nitrogen: 9.5 to 11.5% ( on a dried basis-
Loss on Drying8.0% max
Assay (Titration ex Iodide)% Iodide: 6.6%, max
Formpowder
View more
Polyvinylpyrrolidone-iodine complex is used as a slow iodine release formulation that iodinates lipids and oxidizes cell compounds such as tyrosine side chains. It is utilized to study microbial decontamination of human donor eyes as well as inactivation of infectious hematopoietic necrosis virus. It is also used as a carbon nanotube coating agent to enhance water solubility. It serves as a topical antiseptic in surgery, as an aerosol for skin and mucous membrane infections and is used to prevent infection in wounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only