Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%
Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%
Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%
Thermo Scientific Chemicals

Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 045797.06
also known as 045797-06
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Specifications
CAS63231-67-4
Chemical Name or MaterialSilica gel
EINECS Number231-545-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only