Thermo Scientific Chemicals

Silica gel, HPLC grade, spherical, 5 micron APS, 70 angstroms, 99.99+%

Catalog number: 045809.06
5 g, Each
Thermo Scientific Chemicals

Silica gel, HPLC grade, spherical, 5 micron APS, 70 angstroms, 99.99+%

Catalog number: 045809.06
5 g, Each
Quantity
Catalog number: 045809.06
also known as 045809-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Comment
Pore diameter 6.0-9.0 nm
Comment
Pore volume 0.85-1.05 ml/g
Particle Size
4.30-4.50 Microns
Surface Area
460-540 m²/g
pH
4.0 min

Description

Silica gel, HPLC grade is used in chromatography applications, separation and purification. It is also used in preparative chromatography for HPLC packing, catalyst carriers and resin fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text