Zinc oxide, NanoArc™ ZN-0605
Zinc oxide, NanoArc™ ZN-0605
Zinc oxide, NanoArc™ ZN-0605
Zinc oxide, NanoArc™ ZN-0605
Thermo Scientific Chemicals

Zinc oxide, NanoArc™ ZN-0605

CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045849.22
also known as 045849-22
Price (USD)
33.30
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
33.30
Each
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area12-24m²/g
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis. Used in electronic industry, instrumental industry, manufacture of electrical device, image recorder, rheostat, phosphor. Used as a sun screening agent in cosmetics, antibacterial and health protection antiager; UV protection; Piezoelectrics; High-temperature lubricant in gas turbine engines; Flame retardant; Adsorption; Dental cements; Environmental remediation; Gas sensors; Photocatalytic decontamination; Attenuation of ultraviolet light; Demilitarization of chemical and biological warfare agents; Cosmetics and cosmeceuticals; Electrodes for solar cells; Varistors; Pigments for paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only