Holmium powder, -200 mesh, 99.9% (REO)
Holmium powder, -200 mesh, 99.9% (REO)
Holmium powder, -200 mesh, 99.9% (REO)
Holmium powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Holmium powder, -200 mesh, 99.9% (REO)

CAS: 7440-60-0 | Ho | 164.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 045919.14
also known as 045919-14
Price (USD)
623.65
Special offer
Online exclusive
733.00
Save 109.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
623.65
Special offer
Online exclusive
733.00
Save 109.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only