Dionex™ MetPac™ Oxalic Acid Eluent Concentrate (10X)
Thermo Scientific™

Dionex™ MetPac™ Oxalic Acid Eluent Concentrate (10X)

Choose a high quality eluent concentrate to ensure accurate transitional metals analysis with the Thermo Scientific™ Dionex™ MetPac™ Oxalic Acid Eluent Concentrate.
Have Questions?
Catalog NumberFor Use With (Equipment)
046091IonPac CS5A Column
Catalog number 046091
Price (USD)
-
For Use With (Equipment):
IonPac CS5A Column
Use only high quality, tailor-made eluent concentrates to ensure accurate transitional metals analysis. The Thermo Scientific™ Dionex™ MetPac™ Oxalic Acid Eluent Concentrate is a transitional metal analysis reagent that uses oxalic acid for the reliable separation of lead, copper, cadmium, cobalt, zinc, and nickel on Dionex™ IonPac™ CS5A columns.
Product Detail

The Oxalic Acid Eluent Concentrate is available in 500mL and contains the following;

  • oxalic acid eluent, 10x concentrate

Quality to Trust In

Only high-purity, reagent-grade chemicals and water (18MO-cm), quality-control checked for ionic contamination, are used in Thermo Scientific™ Dionex™ reagents, standards and eluent concentrates.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionMetPac Oxalic Acid Eluent Concentrate (10 X); 500 mL
For Use With (Equipment)IonPac CS5A Column
HazardsIrritant
OperationFor the separation of lead, copper, cadmium, cobalt, zinc, and nickel.
QuantityEach 1
Standard ComponentsOxalic Acid, Tetramethyl Ammonium Hydroxide, Potassium Hydroxide in Deionized Water
Standard Concentration10 X
StorageRoom Temperature
Volume (Metric)500 mL
TypeEluent
Unit SizeEach