Dionex™ IonPac™ CS5A IC Columns
Actual product may vary
Dionex™ IonPac™ CS5A IC Columns
Dionex™ IonPac™ CS5A IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS5A IC Columns

Green features
High capacity ion-exchange column for the fast determination of common transition and lanthanide metals simultaneously in diverse matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
04610450 mm4 mmGuard Column
046100TS
also known as 046100
250 mm4 mmAnalytical Column
052576250 mm2 mmAnalytical Column
05283650 mm2 mmGuard Column
Catalog number 046104
Price (USD)
631.00
Each
Length (Metric):
50 mm
Diameter (Metric):
4 mm
Column Type:
Guard Column
Price (USD)
631.00
Each

Determine common transition and lanthanide metals simultaneously in less than 11 minutes with the Thermo Scientific™ Dionex™ IonPac™ CS5A column. This is a high-resolution column designed for accurate determination of transition and lanthanide metals in a variety of sample matrices. The Dionex IonPac CS5A column, in combination with postcolumn derivitization for visible detection at 530nm, provides a sensitive and selective method for transition metal analysis.

Rapid, Routine Analysis of Transition and Lanthanide Metals in Diverse Samples
  • Sensitive and selective analysis of transition metals in combination with postcolumn derivitization and visible detection at 530 nm.
  • Unique bifunctional resin rapidly provides excellent selectivity for simultaneous determination of common transition and lanthanide metals.
  • Transition metals as well as aluminum are detected following complexation with 4-(2-pyridylazo) resorcinol (PAR).
  • Compatible with up to 50% organic solvents to enhance analyte solubility, modify column selectivity, and simplify column clean-up
Two Eluent Concentrates for Maximum Flexibility
  • Ready-to-dilute, high-purity eluent concentrates are available to easily optimize the Dionex IonPac CS5A column selectivity.
  • The MetPac PDCA eluent uses pyridine-2,6-dicarboxylic acid as a strong complexing agent that separates the metal complexes by anion exchange. The PDCA eluent is best suited for the separation of iron (II) and iron (III), copper, nickel, zinc, cobalt, cadmium, and manganese.
  • The MetPac Oxalic Acid eluent uses oxalic acid as a moderate-strength complexing agent to separate metals by a mixed mode mechanism. The oxalate eluent separates lead, copper, cadmium, cobalt, zinc, and nickel.
Multiple Formats Available for Application Flexibility
  • 4 x 250mm, 2 x 250mm analytical columns
  • 4 x 50mm, 2 x 50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller I.D. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Analysis of Transition and Lanthanide Metals
For Use With (Equipment)Analysis of Transition and Lanthanide Metal
Green FeaturesFSC
Max. Pressure2500 psi (170 bar)
Particle Size9 μm
QuantityEach 1
Solvent CompatibilityUp to 50% Common HPLC Solvents
Stationary PhaseIonPac CG5A
Column TypeGuard Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)50 mm
Unit SizeEach