Platinum crucible with reinforced rim, Top Dia 35mm, Bot Dia21mm, Ht 39mm, Base Thickness 0.3mm, Capacity 25mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum crucible with reinforced rim, Top Dia 35mm, Bot Dia21mm, Ht 39mm, Base Thickness 0.3mm, Capacity 25mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046353.KT
also known as 046353-KT
1 Ea.
Catalog number 046353.KT
also known as 046353-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.