Diaion™ CR11(Na), chelating resin, highly porous type, 0.5 mmole/ml on poly(styrene-divinylbenzene)
Thermo Scientific Chemicals

Diaion™ CR11(Na), chelating resin, highly porous type, 0.5 mmole/ml on poly(styrene-divinylbenzene)

CAS: 74350-44-0
Have Questions?
產品號碼Quantity
046544.A1
亦稱為 046544-A1
1 kg
產品號碼 046544.A1
亦稱為 046544-A1
價格 (TWD)
-
Quantity:
1 kg
Request bulk or custom format
化學識別
CAS74350-44-0
規格Specification Sheet規格表
FormBeads
Appearance (Color)Pale yellow
Assay from Supplier's CofAAdsorption Capacity ≥0.5 mmol/mL.
Assay from Supplier's CofAWater Content 55-65%
Particle Size355-1180?m
Hardness removal in brine, selective transitional metal removal

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only