Thermo Scientific Chemicals

Sepabeads|r SP850, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 45 angstroms, Thermo Scientific Chemicals

Catalog number: 046594.A1
1 kg, Each
Thermo Scientific Chemicals

Sepabeads|r SP850, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 45 angstroms, Thermo Scientific Chemicals

Catalog number: 046594.A1
1 kg, Each
Quantity
Catalog number: 046594.A1
also known as 046594-A1
Price (USD)
Price: 206.00
Online price: 183.65
Your price:
Quantity
-
Appearance (Color)
Pale yellow
Form
Beads
Assay from Supplier's CofA
Water Content 46-52%
Particle Size
250?m
Surface Area
ca. 930m²/g

Description

Amino acid separations, small molecules in medium hydrophobicity in polar solvents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text