Thermo Scientific Chemicals

Indium wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.998+% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号): 046661.G9
25 m, Each
Thermo Scientific Chemicals

Indium wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.998+% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号): 046661.G9
25 m, Each
数量
製品番号(カタログ番号): 046661.G9
または、製品番号046661-G9

化学物質識別子

CAS
7440-74-6
IUPAC Name
indium
Molecular Formula
In
InChI Key
APFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES
[In]
Form
Wire, 1.0mm (0.04in) diameter, annealed.
Appearance (Color)
Silvery
Assay from Supplier's CofA
>99.998% (metals basis)

概要

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text