Non-wetting Pt 5% Au crucible with RF rim, Top Dia 58mm, BotDia 35mm, Ht 64mm, Base Thickness 0.48mm, Capacity 125mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Non-wetting Pt 5% Au crucible with RF rim, Top Dia 58mm, BotDia 35mm, Ht 64mm, Base Thickness 0.48mm, Capacity 125mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046804.KT
also known as 046804-KT
1 Ea.
Catalog number 046804.KT
also known as 046804-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.