Titanium gauze, 60 mesh woven from 0.2mm (0.009in) dia wire
Titanium gauze, 60 mesh woven from 0.2mm (0.009in) dia wire
Titanium gauze, 60 mesh woven from 0.2mm (0.009in) dia wire
Titanium gauze, 60 mesh woven from 0.2mm (0.009in) dia wire
Thermo Scientific Chemicals

Titanium gauze, 60 mesh woven from 0.2mm (0.009in) dia wire

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
75 x 75 mm
150 x 150 mm
Catalog number 046828.HB
also known as 046828-HB
Price (USD)/ Each
653.65
Special offer
Online exclusive
769.00
Save 115.35 (15%)
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
653.65
Special offer
Online exclusive
769.00
Save 115.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
Molecular Weight (g/mol)47.87
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic grey
CommentTitanium: ≥99.5%
FormGauze, 60 mesh woven with 0.009 in. diameter wire
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only