Dicyclopentenyl acrylate
Dicyclopentenyl acrylate
Dicyclopentenyl acrylate
Dicyclopentenyl acrylate
Thermo Scientific Chemicals

Dicyclopentenyl acrylate

CAS: 33791-58-1 | C13H17O2 | 205.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 046864.14
also known as 046864-14
Price (EUR)
82,10
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
82,10
Each
Add to cart
Specifications
CAS33791-58-1
Chemical Name or MaterialDicyclopentenyl acrylate
ColorBrown
Density1.07 g/mL
EINECS Number251-678-1
View more
Dicyclopentenyl acrylate is used in organic synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only