Platinum Standard Dish, Top Dia  38mm, Ht 19mm, Base        Thickness 0.17mm, Capacity 15mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Standard Dish, Top Dia 38mm, Ht 19mm, Base Thickness 0.17mm, Capacity 15mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046878.KT
also known as 046878-KT
1 Ea.
Catalog number 046878.KT
also known as 046878-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.