Bismuth Indium Tin ingot (Field's metal)
Thermo Scientific Chemicals

Bismuth Indium Tin ingot (Field's metal)

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 046895.36
also known as 046895-36
Price (USD)
879.65
Online Exclusive
977.00
Save 97.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
879.65
Online Exclusive
977.00
Save 97.35 (10%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeBi:In:Sn; 32.5:51:16.5 wt%
Chemical Name or MaterialBismuth Indium Tin ingot (Field's metal)
Recommended StorageAmbient temperatures
Solubility InformationInsoluble in water.
TSCAYes
View more

Bismuth Indium Tin ingot (Field's metal) is used for die casting and rapid prototyping. It acts as an alternative to Wood's metal which is an eutectic alloy of 50% bismuth, 26.7% lead, 13.3% tin, and 10% cadmium by weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only