Platinum Standard Dish with reinforced rim, Top Dia  82.5mm, Ht 34mm, Base Thickness 0.25mm, Capacity 150mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Standard Dish with reinforced rim, Top Dia 82.5mm, Ht 34mm, Base Thickness 0.25mm, Capacity 150mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046953.KT
also known as 046953-KT
1 Ea.
Catalog number 046953.KT
also known as 046953-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.