Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100, Thermo Scientific Chemicals
 
Have Questions?
Quantity:
500 mL
Catalog number 047013.AP
also known as 047013-AP
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100, Thermo Scientific Chemicals
Catalog number047013.AP
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
Viscosity?2 cSt at 40?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only