Viscosity standard, Specpure™, nominally 100cSt at 40° and 16.8cSt at 100°
Viscosity standard, Specpure™, nominally 100cSt at 40° and 16.8cSt at 100°
Thermo Scientific Chemicals

Viscosity standard, Specpure™, nominally 100cSt at 40° and 16.8cSt at 100°

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
產品號碼Quantity
047033.AP
亦稱為 047033-AP
500 mL
產品號碼 047033.AP
亦稱為 047033-AP
價格 (TWD)
-
申請報價
Quantity:
500 mL
Request bulk or custom format
規格
CAS8042-47-5
Chemical Name or MaterialViscosity standard
IUPAC Namedisodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
MDL NumberMFCD00084411
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only