Viscosity standard, Specpure™, nominally 120cSt @ 40 and 20cSt @ 100, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Viscosity standard, Specpure™, nominally 120cSt @ 40 and 20cSt @ 100, Thermo Scientific Chemicals
 
Have Questions?
Quantity:
500 mL
Catalog number 047100.AP
also known as 047100-AP
Price (USD)/ Each
115.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
115.00
Add to cart
Viscosity standard, Specpure™, nominally 120cSt @ 40 and 20cSt @ 100, Thermo Scientific Chemicals
Catalog number047100.AP
Price (USD)/ Each
115.00
-
Add to cart
Chemical Identifiers
CAS68649-11-6
SpecificationsSpecification SheetSpecification Sheet
Viscosity?2.4 cSt at 40?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only