Thermo Scientific Chemicals

Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 047178.06
5 g, Each
Thermo Scientific Chemicals

Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 047178.06
5 g, Each
Quantity
Catalog number: 047178.06
also known as 047178-06
Price (USD)
Price: 233.00
Online price: 201.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10138-62-2
IUPAC Name
holmium(3+) trichloride
Molecular Formula
Cl3Ho
InChI Key
PYOOBRULIYNHJR-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ho+3]
Form
Bead
Appearance (Color)
White to pink
Particle Size
-10 Mesh
Total Metal Impurities
0.05% max.

Description

It is mainly used in research. It is used in ceramics, lasers, nuclear industries and in paramagnetic. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text