Thermo Scientific Chemicals

Methanol, LC-MS Grade, 99.8+%, Thermo Scientific Chemicals

4 x 1 L, Each
Thermo Scientific Chemicals

Methanol, LC-MS Grade, 99.8+%, Thermo Scientific Chemicals

4 x 1 L, Each
Quantity
Catalog number: 047192.M1
also known as 047192-M1
Price (USD)
Price: 212.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Appearance (Color)
Clear
Form
Liquid
Color (APHA)
≤5
Assay (unspecified)
LC gradient elution suitability at 254 nm: ≤ 2 mAU
Assay (GC)
≥99.8% (GC-FID)

Description

Solvent for LC-MS analysisMethanol is used as a solvent in Liquid chromatography-mass spectrometry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only