Thermo Scientific Chemicals

Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 047196.03
1 g, Each
Thermo Scientific Chemicals

Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 047196.03
1 g, Each
Quantity
Catalog number: 047196.03
also known as 047196-03
Price (USD)
Price: 185.00
Online price: 160.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14456-51-0
IUPAC Name
thulium(3+) tribromide
Molecular Formula
Br3Tm
InChI Key
HQSWGSFQSCMHFQ-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Tm+3]
Appearance (Color)
White
Form
Bead
Total Rare Earth Oxide Impurities
0.01% max.
Particle Size
-10 Mesh

Description

Thulium(III) bromide is used for development of advanced ceramic discharge lamps1 and metal halide lamp. Reagent for complexation of lanthanide bromides with aluminum bromide. Reactant for preparation of alkali metal thulium bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text