Ytterbium ingot, 99.9% (REO)
Ytterbium ingot, 99.9% (REO)
Thermo Scientific™

Ytterbium ingot, 99.9% (REO)

CAS: 7440-64-4 | Yb | 173.05 g/mol
Have Questions?
Catalog number 047227.KF
also known as 047227-KF
Price (USD)
498.65
Special offer
Online exclusive
587.00
Save 88.35 (15%)
Each
Add to cart
Request bulk or custom format
Price (USD)
498.65
Special offer
Online exclusive
587.00
Save 88.35 (15%)
Each
Add to cart
Specifications
Shelf Life5 Years
CAS7440-64-4
RTECS NumberZG1925000
EINECS Number231-173-2
Molecular FormulaYb
View more