Thermo Scientific Chemicals

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Catalog number: 047240.36
500 g, Each
Thermo Scientific Chemicals

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Catalog number: 047240.36
500 g, Each
Quantity
Catalog number: 047240.36
also known as 047240-36
Price (USD)
Price: 744.00
Online price: 643.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1306-38-3
IUPAC Name
dioxocerium
Molecular Formula
CeO2
InChI Key
CETPSERCERDGAM-UHFFFAOYSA-N
SMILES
O=[Ce]=O
Concentration (weight/weight)
20% min

Description

The addition of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures. It is used as an additive in glass industry. Cerium dioxide is mainly used as such or in formulation as a high precision polishing agent for glass products, as catalyst or catalyst support. Polishing and decolorizing glass; opacifier in vitreous enamels and photochromic glasses; heat-resistant alloy coatings; as a cracking catalyst; as a catalyst for automobile emission control; in ceramic coatings; in phosphors; in cathodes; in capacitors; in semiconductors; in refractory oxides; gemstone polishing, as raw material for the production of glass and ceramics as well as additives in paints and varnishes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text