Product Image
Thermo Scientific Chemicals

Silicon, AAS standard solution, Specpure™ Si 1000μg/mL

 
Have Questions?
產品號碼Quantity
047277.AE
亦稱為 047277-AE
100 mL
產品號碼 047277.AE
亦稱為 047277-AE
價格 (TWD)
-
Quantity:
100 mL
Request bulk or custom format
規格
Chemical Name or MaterialSilicon, AAS standard solution
MDL NumberMFCD00085311
Name NoteSi 1000μg/mL
Physical FormLiquid
Quantity100 mL
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only