Boron carbide, 99+%
Thermo Scientific Chemicals

Boron carbide, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047378.30
also known as 047378-30
Price (EUR)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS12069-32-8
IUPAC Name2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
Molecular FormulaCB4
InChI KeyINAHAJYZKVIDIZ-UHFFFAOYSA-N
SMILESB12B3B4B1C234
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey or black
FormPowder, -325 mesh
Assay from Supplier's CofABoron Carbide: ≥99.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.