DRI™ Endocrine Calibrator Sets
Actual product may vary
DRI™ Endocrine Calibrator Sets
DRI™ Endocrine Calibrator Sets
Thermo Scientific™

DRI™ Endocrine Calibrator Sets

Achieve excellent workflow management, cost savings and superior performance with these endocrine calibrator sets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
04766 x 2 mL
07385 x 2 mL
Catalog number 0476
Price (CLP)
-
Quantity:
6 x 2 mL
Achieve excellent workflow management, cost savings and superior performance with Thermo Scientific™ DRI™ Endocrine Calibrator Sets. Packaged in convenient sizes with applications on a multitude of clinical chemistry analyzers, Endocrine Assays T4 and T Uptake are available in both Thermo Scientific™ CEDIA™ and DRI formats.
  • Packaged in convenient sizes
  • Applications on a multitude of clinical chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionTotal Thyroxine (T4) Calibrator Kit
Quantity6 x 2 mL
TypeCalibrator
Detectable AnalytesNegative Calibrator, Thyroxine, Thyroxine
Sample TypePlasma, Serum
Unit SizeEach
Contents & Storage
2°C to 8°C