Dionex™ ASE™ 150/200/350 Collection Vials
Actual product may vary
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Dionex™ ASE™ 150/200/350 Collection Vials
Thermo Scientific™

Dionex™ ASE™ 150/200/350 Collection Vials

Collect extract from Thermo Scientific™ Dionex™ ASE™ 150/200/350 Accelerated Solvent Extractors using these 40mL and 60mL collection vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionColorVolume (Metric)For Use With (Equipment)
048784Collection VialClear60 mLDionex ASE 150, 350, and 200
048780Collection VialAmber40 mLDionex ASE 200
048783Collection VialClear40 mLDionex ASE 200
048781Collection VialAmber60 mLDionex ASE 150, 350, and 200
055442
also known as 55442
Graduated concentration vial40 mLDionex ASE 200
055444Spacer, graduated vialDionex ASE 200, Graduated Vials
Catalog number 048784
Price (USD)
211.00
Case of 72
Description:
Collection Vial
Color:
Clear
Volume (Metric):
60 mL
For Use With (Equipment):
Dionex ASE 150, 350, and 200
Price (USD)
211.00
Case of 72
Collect solvent and analytes extracted from Thermo Scientific™ Dionex™ ASE™ 150/200/350 Accelerated Solvent Extractors using these collection vials. Placed in the vial tray at the lower right of the system, these vials are available in 40 and 60mL clear and amber glass, and are complete with caps and solvent-resistant (TFE) septa.
Compatibility
  • 40mL and 40mL graduated vials are only compatible with Dionex ASE 200 systems.
  • 40mL collection vessels are not compatible with Dionex ASE 150 or 350 systems.
  • 60mL collection vials require an adaptor for use with Dionex ASE 150 systems, and are not compatible with Dionex ASE 100 systems.

See Dionex™ ASE™ Collection Bottles and Lids to order 250mL collection bottles for larger volume extraction collection.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ColorClear
DescriptionCollection Vial
QuantityCase of 72
Volume (Metric)60 mL
For Use With (Equipment)Dionex ASE 150, 350, and 200
TypeCollection Vial
Unit SizeCase of 72