Dionex™ ASE™ Extraction Cell O-Ring
Actual product may vary
Dionex™ ASE™ Extraction Cell O-Ring
Dionex™ ASE™ Extraction Cell O-Ring
Thermo Scientific™
Dionex™ ASE™ Extraction Cell O-Ring
Ensure a proper seal and maximize extraction efficiency with periodic replacement of these Thermo Scientific™ Dionex™ ASE™ Extraction Cell O-Rings for Dionex ASE Accelerated Solvent Extractor cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial
049457Teflon
056325Viton
Catalog number 049457
Price (MXN)
-
Material:
Teflon
Ensure a proper seal and maximize extraction efficiency with periodic replacement of these Thermo Scientific™ Dionex™ ASE™ Extraction Cell O-Rings for Dionex ASE Accelerated Solvent Extractor cells. Choose from PTFE or Viton™ O-rings to suit your requirements.
Dionex ASE systems are used to reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices.
Ordering Information:
Package of 50
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
MaterialTeflon
Quantity50
For Use With (Equipment)Dionex ASE 100, 150, 200, 300 or 350 for Outside Extraction Cell Caps
TypeO-ring
Unit SizePack of 50