Thermo Scientific™
Capitol Vial Flip-Top Vials with Lock Seal
Related applications:
Tubes and Vials
Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal are similar to standard flip-top vials except they contain an additional lock-seal tab.
Back to top
  • 100% FDA approved food grade polypropylene
  • Sterile, airtight and leak-resistant construction
  • Permanent hinged lid allows easy one-handed opening of vial
  • Shoulderless, linerless containers
  • Available in capacities ranging from 1.5 to 10 oz. (44.3 to 296mL)
Recommended for:
Biomaterial storage, biopsy transport, DNA storage, dairy testing, diagnostic product packaging, environmental sampling, water testing
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.